Anti-PPARA Mouse Monoclonal Antibody [clone: 1E8]

Lieferant: OriGene
CF503540
ORIGCF503540EA 833 EUR
ORIGCF503540
Anti-PPARA Mouse Monoclonal Antibody [clone: 1E8]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100

Type: Primary
Antigen: PPARA
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR