Anti-MTFMT Mouse Monoclonal Antibody [clone: 1A10]

Lieferant: OriGene
CF503564
ORIGCF503564EA 833 EUR
ORIGCF503564
Anti-MTFMT Mouse Monoclonal Antibody [clone: 1A10]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: MTFMT
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR