Anti-EIF2B3 Mouse Monoclonal Antibody [clone: 2D2]

Lieferant: OriGene
CF503568
ORIGCF503568EA 833 EUR
ORIGCF503568
Anti-EIF2B3 Mouse Monoclonal Antibody [clone: 2D2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: EIF2B3
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR