Anti-CDKN2B Mouse Monoclonal Antibody [clone: 8H4]

Lieferant: OriGene
CF505208
ORIGCF505208EA 833 EUR
ORIGCF505208
Anti-CDKN2B Mouse Monoclonal Antibody [clone: 8H4]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CDKN2B
Clonality: Monoclonal
Clone: 8H4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR