Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 6C5]

Lieferant: OriGene
CF505826
ORIGCF505826EA 833 EUR
ORIGCF505826
Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 6C5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PIK3R5
Clonality: Monoclonal
Clone: 6C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR