Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 3H2]

Lieferant: OriGene
CF800059
ORIGCF800059EA 833 EUR
ORIGCF800059
Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 3H2]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: UBE2E3
Clonality: Monoclonal
Clone: 3H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR