Anti-NME1 Mouse Monoclonal Antibody [clone: 18C12]

Lieferant: OriGene
CF801300
ORIGCF801300EA 833 EUR
ORIGCF801300
Anti-NME1 Mouse Monoclonal Antibody [clone: 18C12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NME1
Clonality: Monoclonal
Clone: 18C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR