Anti-KI67 Rat Monoclonal Antibody [clone: 3B6]

Lieferant: OriGene
CF801343
ORIGCF801343EA 833 EUR
ORIGCF801343
Anti-KI67 Rat Monoclonal Antibody [clone: 3B6]
Antikörper
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: KI67
Clonality: Monoclonal
Clone: 3B6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR