Anti-NME1 Mouse Monoclonal Antibody [clone: 2H10]

Lieferant: OriGene
CF801361
ORIGCF801361EA 833 EUR
ORIGCF801361
Anti-NME1 Mouse Monoclonal Antibody [clone: 2H10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NME1
Clonality: Monoclonal
Clone: 2H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR