Anti-PGR Mouse Monoclonal Antibody [clone: 18B12]

Lieferant: OriGene
CF802718
ORIGCF802718EA 833 EUR
ORIGCF802718
Anti-PGR Mouse Monoclonal Antibody [clone: 18B12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 18B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR