Anti-FGFR1 Mouse Monoclonal Antibody [clone: 5D5]

Lieferant: OriGene
CF803096
ORIGCF803096EA 833 EUR
ORIGCF803096
Anti-FGFR1 Mouse Monoclonal Antibody [clone: 5D5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FGFR1
Clonality: Monoclonal
Clone: 5D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR