Anti-RIPK3 Mouse Monoclonal Antibody [clone: 2C11]

Lieferant: OriGene
CF803101
ORIGCF803101EA 833 EUR
ORIGCF803101
Anti-RIPK3 Mouse Monoclonal Antibody [clone: 2C11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RIPK3
Clonality: Monoclonal
Clone: 2C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR