Anti-VWF Mouse Monoclonal Antibody [clone: 2F2]

Lieferant: OriGene
CF803265
ORIGCF803265EA 833 EUR
ORIGCF803265
Anti-VWF Mouse Monoclonal Antibody [clone: 2F2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: 2F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR