Anti-PDE1A Mouse Monoclonal Antibody [clone: 7D5]

Lieferant: OriGene
CF803629
ORIGCF803629EA 833 EUR
ORIGCF803629
Anti-PDE1A Mouse Monoclonal Antibody [clone: 7D5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PDE1A
Clonality: Monoclonal
Clone: 7D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR