Anti-ZNF286A Mouse Monoclonal Antibody [clone: 6A5]

Lieferant: OriGene
CF803642
ORIGCF803642EA 833 EUR
ORIGCF803642
Anti-ZNF286A Mouse Monoclonal Antibody [clone: 6A5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ZNF286A
Clonality: Monoclonal
Clone: 6A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR