Anti-6xHis Monoclonal Antibody

Lieferant: OriGene
TA100013
ORIGTA100013EA 452 EUR
ORIGTA100013
Anti-6xHis Monoclonal Antibody
Antikörper
Type: Tag
Antigen: 6XHis
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR