Anti-ZsGreen1 IgG1 Monoclonal Antibody (Magnetic beads) [clone: OTI2C2 (formerly 2C2)]
Lieferant: OriGene
TA183023
ORIGTA183023EA 1080 EUR
ORIGTA183023
Anti-ZsGreen1 IgG1 Monoclonal Antibody (Magnetic beads) [clone: OTI2C2 (formerly 2C2)]
Antikörper
Type: Tag
Antigen: ZsGreen1
Clonality: Monoclonal
Clone: OTI2C2 (formerly 2C2)
Conjugation: Magnetic beads
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen