Anti-ZsYellow1 IgG1 Monoclonal Antibody (Magnetic beads) [clone: OTI3C4 (formerly 3C4)]
Lieferant: OriGene
TA183024
ORIGTA183024EA 1080 EUR
ORIGTA183024
Anti-ZsYellow1 IgG1 Monoclonal Antibody (Magnetic beads) [clone: OTI3C4 (formerly 3C4)]
Antikörper
Type: Tag
Antigen: ZsYellow1
Clonality: Monoclonal
Clone: OTI3C4 (formerly 3C4)
Conjugation: Magnetic beads
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen