Anti-mCherry IgG1 Monoclonal Antibody (Magnetic beads) [clone: OTI10G6 (formerly 10G6)]

Lieferant: OriGene
TA183030
ORIGTA183030EA 1080 EUR
ORIGTA183030
Anti-mCherry IgG1 Monoclonal Antibody (Magnetic beads) [clone: OTI10G6 (formerly 10G6)]
Antikörper
Type: Tag
Antigen: mCherry
Clonality: Monoclonal
Clone: OTI10G6 (formerly 10G6)
Conjugation: Magnetic beads
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR