Anti-mRaspberry IgG1 Monoclonal Antibody (Magnetic beads) [clone: OTI2A6 (formerly 2A6)]

Lieferant: OriGene
TA183031
ORIGTA183031EA 1080 EUR
ORIGTA183031
Anti-mRaspberry IgG1 Monoclonal Antibody (Magnetic beads) [clone: OTI2A6 (formerly 2A6)]
Antikörper
Type: Tag
Antigen: mRaspberry
Clonality: Monoclonal
Clone: OTI2A6 (formerly 2A6)
Conjugation: Magnetic beads
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR