Anti-E2-Crimson IgG2a Monoclonal Antibody (Magnetic beads) [clone: OTI9B3 (formerly 9B3)]
Lieferant: OriGene
TA183040
ORIGTA183040EA 1080 EUR
ORIGTA183040
Anti-E2-Crimson IgG2a Monoclonal Antibody (Magnetic beads) [clone: OTI9B3 (formerly 9B3)]
Antikörper
Type: Tag
Antigen: E2-Crimson
Clonality: Monoclonal
Clone: OTI9B3 (formerly 9B3)
Conjugation: Magnetic beads
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen