Anti-Dendra2 IgG2b Monoclonal Antibody (Magnetic beads) [clone: OTI1G6 (formerly 1G6)]
Lieferant: OriGene
TA183044
ORIGTA183044EA 1080 EUR
ORIGTA183044
Anti-Dendra2 IgG2b Monoclonal Antibody (Magnetic beads) [clone: OTI1G6 (formerly 1G6)]
Antikörper
Type: Tag
Antigen: Dendra2
Clonality: Monoclonal
Clone: OTI1G6 (formerly 1G6)
Conjugation: Magnetic beads
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen