Anti-LAT Goat Polyclonal Antibody

Lieferant: OriGene
TA302672
ORIGTA302672EA 785 EUR
ORIGTA302672
Anti-LAT Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: LAT (linker for activation of T cells)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR