Anti-ERLIN2 Goat Polyclonal Antibody

Lieferant: OriGene
TA303052
ORIGTA303052EA 785 EUR
ORIGTA303052
Anti-ERLIN2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:2000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: ERLIN2 (ER lipid raft associated 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR