Anti-COG7 Goat Antibody

Lieferant: OriGene
TA305712
ORIGTA305712EA 785 EUR
ORIGTA305712
Anti-COG7 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: COG7 (component of oligomeric golgi complex 7)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR