Anti-RRM2B Rabbit Polyclonal Antibody

Lieferant: OriGene
TA306062
ORIGTA306062EA 792 EUR
ORIGTA306062
Anti-RRM2B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: RRM2B (ribonucleotide reductase M2 B (TP53 inducible))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR