Anti-RNASEH2A Rabbit Polyclonal Antibody

Lieferant: OriGene
TA306706
ORIGTA306706EA 792 EUR
ORIGTA306706
Anti-RNASEH2A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 2 ?g/ml; Immunofluorescence: 4 ?g/ml

Type: Primary
Antigen: RNASEH2A (ribonuclease H2, subunit A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR