Anti-MZB1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA308110
ORIGTA308110EA 868 EUR
ORIGTA308110
Anti-MZB1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: MZB1 (marginal zone B and B1 cell-specific protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR