Anti-PRPSAP2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA308166
ORIGTA308166EA 868 EUR
ORIGTA308166
Anti-PRPSAP2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: PRPSAP2 (phosphoribosyl pyrophosphate synthetase-associated protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR