Anti-RALB Rabbit Polyclonal Antibody

Lieferant: OriGene
TA308189
ORIGTA308189EA 868 EUR
ORIGTA308189
Anti-RALB Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: RALB (v-ral simian leukemia viral oncogene homolog B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR