Anti-HS1BP3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA308194
ORIGTA308194EA 868 EUR
ORIGTA308194
Anti-HS1BP3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:5000-1:20000

Type: Primary
Antigen: HS1BP3 (HCLS1 binding protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR