Anti-ANXA1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA308701
ORIGTA308701EA 868 EUR
ORIGTA308701
Anti-ANXA1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:1000-1:10000

Type: Primary
Antigen: ANXA1 (annexin A1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR