Anti-GIMAP8 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA309055
ORIGTA309055EA 868 EUR
ORIGTA309055
Anti-GIMAP8 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: GIMAP8 (GTPase, IMAP family member 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR