Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: 8D3]

Lieferant: OriGene
TA309394
ORIGTA309394EA 528 EUR
ORIGTA309394
Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: 8D3]
Antikörper
  • Recommended Dilutions:

Type: Primary
Antigen: HSP90AB1 (heat shock protein 90kDa alpha (cytosolic), class B member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR