Anti-DLG4 Mouse Monoclonal Antibody [clone: S28-86]

Lieferant: OriGene
TA309458
ORIGTA309458EA 792 EUR
ORIGTA309458
Anti-DLG4 Mouse Monoclonal Antibody [clone: S28-86]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: DLG4 (discs, large homolog 4 (Drosophila))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR