Anti-LRRK2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA309736
ORIGTA309736EA 847 EUR
ORIGTA309736
Anti-LRRK2 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions:

Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR