Anti-NAPSA Mouse Monoclonal Antibody [clone: TMU-Ad02]

Lieferant: OriGene
TA309758
ORIGTA309758EA 847 EUR
ORIGTA309758
Anti-NAPSA Mouse Monoclonal Antibody [clone: TMU-Ad02]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: NAPSA (napsin A aspartic peptidase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR