Anti-NKX6-1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA309767
ORIGTA309767EA 944 EUR
ORIGTA309767
Anti-NKX6-1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: NKX6-1 (NK6 homeobox 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR