Anti-RETN Rabbit Polyclonal Antibody

Lieferant: OriGene
TA309836
ORIGTA309836EA 589 EUR
ORIGTA309836
Anti-RETN Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: RETN (resistin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR