Anti-SAP30L Rabbit Polyclonal Antibody

Lieferant: OriGene
TA309844
ORIGTA309844EA 558 EUR
ORIGTA309844
Anti-SAP30L Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: SAP30L (SAP30-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR