Anti-FOXO4 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA309984
ORIGTA309984EA 785 EUR
ORIGTA309984
Anti-FOXO4 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: FOXO4 (forkhead box O4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR