Anti-MAP2K6 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA309987
ORIGTA309987EA 785 EUR
ORIGTA309987
Anti-MAP2K6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: MAP2K6 (mitogen-activated protein kinase kinase 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR