Anti-ADD1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA310000
ORIGTA310000EA 785 EUR
ORIGTA310000
Anti-ADD1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: ADD1 (adducin 1 (alpha))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR