Anti-BRCA1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA310042
ORIGTA310042EA 785 EUR
ORIGTA310042
Anti-BRCA1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR