Anti-DOK2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA310073
ORIGTA310073EA 785 EUR
ORIGTA310073
Anti-DOK2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: DOK2 (docking protein 2, 56kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR