Anti-WNT5B Rabbit Monoclonal Antibody [clone: EPR7135(B)]

Lieferant: OriGene
TA310943
ORIGTA310943EA 778 EUR
ORIGTA310943
Anti-WNT5B Rabbit Monoclonal Antibody [clone: EPR7135(B)]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Flow Cytometry: 1:10-1:100

Type: Primary
Antigen: WNT5B (wingless-type MMTV integration site family, member 5B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR