Anti-FANCL Goat Antibody

Lieferant: OriGene
TA311186
ORIGTA311186EA 785 EUR
ORIGTA311186
Anti-FANCL Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: FANCL (Fanconi anemia, complementation group L)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR