Anti-B2M Goat Antibody

Lieferant: OriGene
TA311281
ORIGTA311281EA 785 EUR
ORIGTA311281
Anti-B2M Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: B2M (beta-2-microglobulin)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR