Anti-HTR1A Goat Antibody

Lieferant: OriGene
TA311385
ORIGTA311385EA 785 EUR
ORIGTA311385
Anti-HTR1A Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: HTR1A (5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR