Anti-TNFRSF1B Goat Antibody

Lieferant: OriGene
TA311396
ORIGTA311396EA 785 EUR
ORIGTA311396
Anti-TNFRSF1B Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.2-0.6?g/ml

Type: Primary
Antigen: TNFRSF1B (tumor necrosis factor receptor superfamily, member 1B)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR